(±)-α-Lipoic acid 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

138720050 138720250 138721000
ACRO138720050EA 642 DKK
ACRO138720050 ACRO138720250 ACRO138721000
(±)-α-Lipoic acid 98+%
(±)-α-Lipoic acid
Formel: C₈H₁₄O₂S₂
Kogepunkt: 160…165 °C (760 mmHg)
Smeltepunkt: 60…63 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005474
CAS-nummer: 1077-28-7
EINECS: 214-071-2

Bestil nu

Specifikationer Testresultater

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
UV spectrum Complies
Melting point 60 to 63°C
Loss on drying ≤1 %
Sulfated ash ≤0.1 %
Solubility (2 % in 10 % NaOH) Clear yellow
UV spectrophotometry 98.0 to 102.0 % (on dry substance)
Chloride (Cl) ≤0.071 %
Sulfate (SO4) ≤0.192 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR