(±)-Dimethyl acetylsuccinate ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Dimethyl acetylsuccinate
A1143-25G
A1143-500G
TCIAA1143-25GEA
132
DKK
TCIAA1143-25G
TCIAA1143-500G
(±)-Dimethyl acetylsuccinate ≥96.0% (by GC)
(±)-Dimethyl acetylsuccinate
Formel:
C₈H₁₂O₅ Molekylvægt: 188.18 g/mol Kogepunkt: 200°C |
MDL Number:
MFCD00008448 CAS-nummer: 10420-33-4 EINECS: 233-897-4 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...