(±)-Dimethyl acetylsuccinate 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Dimethyl acetylsuccinate
B25423.22
B25423.36
B25423.22EA
513
DKK
B25423.22
B25423.36
(±)-Dimethyl acetylsuccinate 96%
(±)-Dimethyl acetylsuccinate
Purity: 96%
CAS No.: 10420-33-4
Molecular Formula: C8H12O5
Formula Weight: 188.18
Flash Point:
Formel:
C₈H₁₂O₅ Molekylvægt: 188.18 g/mol Kogepunkt: 129…134 °C (12 mmHg) Smeltepunkt: 30…33 °C Massefylde: 1.160 Flammepunkt: >110 °C (230 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008448 CAS-nummer: 10420-33-4 EINECS: 233-897-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...