2-Fluoro-L-phenylalanine ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: H-Phe(2-F)-OH

F0273-1G F0273-100MG
TCIAF0273-1GEA 559 DKK
TCIAF0273-1G TCIAF0273-100MG
2-Fluoro-L-phenylalanine ≥96.0% (by titrimetric analysis)
2-Fluoro-L-phenylalanine
Formel: C₉H₁₀FNO₂
MDL Number: MFCD00066448
CAS-nummer: 19883-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=170b731d94223eaa6cb34fc96e7d08aa535eb24d&vid=170b731d94223eaa6cb34fc96e7d08aa535eb24d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D -12.0 to -18.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR