Benzylchlorformiat 30 - 35% in toluene

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C0176-25ML C0176-500ML
TCIAC0176-25MLEA 181 DKK
TCIAC0176-25ML TCIAC0176-500ML
Benzylchlorformiat 30 - 35% in toluene
Benzylchlorformiat
Formel: C₈H₇ClO₂
MDL Number: MFCD00000640
CAS-nummer: 501-53-1
EINECS: 207-925-0
UN nummer: 2920
ADR: 8,II
Merck Index: 14,01801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b879fd1bd831dfd961505b4100b602c77d5b2ec&vid=6a56d29c8f918187e1cda40b22078ae254e5fdc1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 30.0 to 35.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR