trans-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: trans-4-(tert-Butyl)cyclohexanecarboxylic acid

B3037-5G B3037-1G
TCIAB3037-5GEA 1890 DKK
TCIAB3037-5G TCIAB3037-1G
trans-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-tert-Butylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
MDL Number: MFCD00466237
CAS-nummer: 943-29-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad4f72939eb21b99eb3c8fcb35a9fe5650e5960&vid=4ad4f72939eb21b99eb3c8fcb35a9fe5650e5960&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 174.0 to 178.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR