1-Bromo-4-(trifluoromethylthio)benzene ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 97%
CAS No.: 333-47-1
Molecular Formula: C7H4BrF3S
Formula Weight: 257.07
Flash Point: 83°(181°F)
Refractive Index: 1.5170
Boiling Point: 199-200°
Beilstein Reference: 2361926
MDL No.: MFCD00040840
Formel:
C₇H₄BrF₃S Kogepunkt: 199…200 °C Massefylde: 1,710 Flammepunkt: 83 °C (181 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00040840 CAS-nummer: 333-47-1 |