3-Amino-2-methylbenzotrifluoride ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methyl-3-(trifluoromethyl)aniline

M1328-1G M1328-5G
TCIAM1328-1GEA 329 DKK
TCIAM1328-1G TCIAM1328-5G
3-Amino-2-methylbenzotrifluoride ≥99.0% (by GC)
3-Amino-2-methylbenzotrifluoride
Formel: C₈H₈F₃N
Molekylvægt: 175.16 g/mol
MDL Number: MFCD00153216
CAS-nummer: 54396-44-0
EINECS: 259-145-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3230f25a759d3a2038f31ae65b552cdcef59cdaf&vid=3230f25a759d3a2038f31ae65b552cdcef59cdaf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 99.0 %
Melting point 38.0 to 41.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR