Thiazole-2-carbaldehyde ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

F0303-1G F0303-5G
TCIAF0303-1GEA 148 DKK
TCIAF0303-1G TCIAF0303-5G
Thiazole-2-carbaldehyde ≥97.0% (by GC)
Thiazole-2-carbaldehyde
Formel: C₄H₃NOS
Kogepunkt: 70°C
Flammepunkt: 68°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00142924
CAS-nummer: 10200-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28c37afebbd35f9306a7534e890a1c99e9b8fa75&vid=28c37afebbd35f9306a7534e890a1c99e9b8fa75&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR