1,4-Bis(trifluoromethyl)-2-nitrobenzene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Nitro-1,4-bis(trifluoromethyl)benzene, 1-Nitro-2,5-bis(trifluoromethyl)benzene
B2546-25G
B2546-5G
TCIAB2546-25GEA
952
DKK
TCIAB2546-25G
TCIAB2546-5G
1,4-Bis(trifluoromethyl)-2-nitrobenzene ≥98.0% (by GC)
1,4-Bis(trifluoromethyl)-2-nitrobenzene
Formel:
C₈H₃F₆NO₂ Kogepunkt: 90°C |
MDL Number:
MFCD00236674 CAS-nummer: 320-88-7 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...