2-Chloro-N,N-diethylacetamide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D0483-25G
D0483-500G
TCIAD0483-25GEA
312
DKK
TCIAD0483-25G
TCIAD0483-500G
2-Chloro-N,N-diethylacetamide ≥98.0% (by GC)
2-Chloro-N,N-diethylacetamide
Formel:
C₆H₁₂ClNO Kogepunkt: 150°C |
MDL Number:
MFCD00000928 CAS-nummer: 2315-36-8 EINECS: 219-019-2 UN nummer: 2810 ADR: 6.1,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...