2(5H)-Furanon ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C1078-25ML C1078-5ML
TCIAC1078-25MLEA 2480 DKK
TCIAC1078-25ML TCIAC1078-5ML
2(5H)-Furanon ≥93.0% (by GC)
2(5H)-Furanon
Formel: C₄H₄O₂
Kogepunkt: 214°C
Smeltepunkt: 5°C
Flammepunkt: 101°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005376
CAS-nummer: 497-23-4
EINECS: 207-839-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb0f46e14ced0838bc8d96990107f3ba86e31171&vid=eb0f46e14ced0838bc8d96990107f3ba86e31171&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR