2,2'-Dithiodi(benzoesyre) ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,2'-Dithiosalicylic acid, 2,2'-Dithiodibenzoic acid

D1437-500G D1437-25G D1437-100G
TCIAD1437-500GEA 5220 DKK
TCIAD1437-500G TCIAD1437-25G TCIAD1437-100G
2,2'-Dithiodi(benzoesyre) ≥96.0% (by HPLC, titration analysis)
2,2'-Dithiodi(benzoesyre)
Formel: C₁₄H₁₀O₄S₂
MDL Number: MFCD00002465
CAS-nummer: 119-80-2
EINECS: 204-352-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4df5a578e95ccf1d462a293fec270e1371917e99&vid=4df5a578e95ccf1d462a293fec270e1371917e99&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR