Bis(cyclopentadienyl)titaniumdichlorid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

T0616-25G T0616-5G
TCIAT0616-25GEA 657 DKK
TCIAT0616-25G TCIAT0616-5G
Bis(cyclopentadienyl)titaniumdichlorid ≥98.0% (by titrimetric analysis)
Bis(cyclopentadienyl)titaniumdichlorid
Formel: C₁₀H₁₀Cl₂Ti
Molekylvægt: 248.97 g/mol
MDL Number: MFCD00003723
CAS-nummer: 1271-19-8
EINECS: 215-035-9
Merck Index: 14,09482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41fe014061709555d928ec48876f10c1e4b13bba&vid=41fe014061709555d928ec48876f10c1e4b13bba&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in THF very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR