2,4-Quinolindiol ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D1753-25G
D1753-500G
TCIAD1753-25GEA
501
DKK
TCIAD1753-25G
TCIAD1753-500G
2,4-Quinolindiol ≥97.0% (by GC, titration analysis)
2,4-Quinolindiol
Formel:
C₉H₇NO₂ Kogepunkt: 375 °C (1013 hPa) Smeltepunkt: 159 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006744 CAS-nummer: 86-95-3 EINECS: 201-711-0 |
|
Specifikationer Testresultater
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...