5-Nitro-m-xylen ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 1,3-Dimethyl-5-nitrobenzene
D1086-25G D1086-5G
TCIAD1086-25GEA 1180 DKK
TCIAD1086-25G TCIAD1086-5G
5-Nitro-m-xylen ≥98.0% (by GC)
5-Nitro-m-xylen
Formel: C₈H₉NO₂
Molekylvægt: 151.16 g/mol
Kogepunkt: 273°C
Smeltepunkt: 70
MDL Number: MFCD00007269
CAS-nummer: 99-12-7
EINECS: 202-732-8
UN nummer: 3447
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d492f9bc65ab7b7ddb13e01f68150be6df02fe33&vid=d492f9bc65ab7b7ddb13e01f68150be6df02fe33&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in Methanol very faint turbidity