1-Phenyl-1-decanon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Decanophenone, Decyl phenyl ketone, Nonyl phenyl ketone
D0026-25G
D0026-500G
TCIAD0026-25GEA
370
DKK
TCIAD0026-25G
TCIAD0026-500G
1-Phenyl-1-decanon ≥98.0% (by GC)
1-Phenyl-1-decanon
Formel:
C₆H₅C(O)(CH₂)₈CH₃ Kogepunkt: 187°C Smeltepunkt: 35 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00009582 CAS-nummer: 6048-82-4 EINECS: 227-946-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 35.0 to 37.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...