2-(4-Chlorbenzoyl)benzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C1074-25G
C1074-500G
TCIAC1074-25GEA
132
DKK
TCIAC1074-25G
TCIAC1074-500G
2-(4-Chlorbenzoyl)benzoesyre ≥98.0% (by GC, titration analysis)
2-(4-Chlorbenzoyl)benzoesyre
Formel:
C₁₄H₉ClO₃ |
MDL Number:
MFCD00002474 CAS-nummer: 85-56-3 EINECS: 201-615-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...