Diethylcyclopropan-1,1-dicarboxylat ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Diethyl 1,1-cyclopropanedicarboxylate, Diethyl 1,1-cyclopropane dicarboxyliate

C1341-25G C1341-5G
TCIAC1341-25GEA 706 DKK
TCIAC1341-25G TCIAC1341-5G
Diethylcyclopropan-1,1-dicarboxylat ≥96.0% (by GC)
Diethylcyclopropan-1,1-dicarboxylat
Formel: C₉H₁₄O₄
Kogepunkt: 215°C
Flammepunkt: 86°C
MDL Number: MFCD00001283
CAS-nummer: 1559-02-0
EINECS: 216-321-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7518083c2dc081aa16adbbde4a058b70629df39f&vid=7518083c2dc081aa16adbbde4a058b70629df39f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR