4-(Trifluormethyl)-2,3,5,6-tetrafluorthiophenol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1542-1G T1542-5G
TCIAT1542-1GEA 337 DKK
TCIAT1542-1G TCIAT1542-5G
4-(Trifluormethyl)-2,3,5,6-tetrafluorthiophenol ≥97.0% (by GC, titration analysis)
4-(Trifluormethyl)-2,3,5,6-tetrafluorthiophenol
Formel: C₇HF₇S
Kogepunkt: 169°C
MDL Number: MFCD00191594
CAS-nummer: 651-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d054d22eefdc10b0bbc4ff3593bc4e5eb1fed473&vid=d054d22eefdc10b0bbc4ff3593bc4e5eb1fed473&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR