n-Pentadecan ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
N-Pentadecane, Pentadecane
P0606-25ML
P0606-500ML
P0606-100ML
TCIAP0606-25MLEA
238
DKK
TCIAP0606-25ML
TCIAP0606-500ML
TCIAP0606-100ML
n-Pentadecan ≥99.0% (by GC)
n-Pentadecane
Formel:
H₃C(CH₂)₁₃CH₃ Kogepunkt: 270°C Smeltepunkt: 13…15 °C Massefylde: 0,768 g/cm³ (20 °C) Flammepunkt: 120°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008990 CAS-nummer: 629-62-9 EINECS: 211-098-1 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...