n-Octadecan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Octadecane
O0003-25G
O0003-250G
TCIAO0003-25GEA
214
DKK
TCIAO0003-25G
TCIAO0003-250G
n-Octadecan ≥98.0% (by GC)
n-Octadecane
Formel:
H₃C(CH₂)₁₆CH₃ Kogepunkt: 316,1 °C (1013 hPa) Smeltepunkt: 26…29 °C Massefylde: 0,7765 g/cm³ (20 °C) Flammepunkt: 165 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009007 CAS-nummer: 593-45-3 EINECS: 209-790-3 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...