8-Chlor-1-octanol ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 8-Chloro-1-n-octanol

C2448-25ML C2448-5ML
TCIAC2448-25MLEA 805 DKK
TCIAC2448-25ML TCIAC2448-5ML
8-Chlor-1-octanol ≥97.0% (by GC)
8-Chloro-1-octanol
Formel: C₈H₁₇ClO
Kogepunkt: 130°C
Smeltepunkt: –3 °C
Massefylde: 0,976 g/cm³ (25 °C)
MDL Number: MFCD00040005
CAS-nummer: 23144-52-7
EINECS: 245-451-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bfa049b848fa522b52a7fbf2ec0e230c09e8555&vid=9d766fe4391d1a73f00c8b1b6e8e2f09ba77c1f5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR