Amantadin, HCl ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride
A0588-25G A0588-250G
TCIAA0588-25GEA 329 DKK
TCIAA0588-25G TCIAA0588-250G
Amantadin, HCl ≥99.0% (by titrimetric analysis)
Amantadin, HCl
Formel: C₁₀H₁₈ClN
Molekylvægt: 187.71 g/mol
Smeltepunkt: 360
MDL Number: MFCD00074723
CAS-nummer: 665-66-7
EINECS: 211-560-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5f5450ef5ca791049eec5f7d1d136a1f7195123&vid=a5f5450ef5ca791049eec5f7d1d136a1f7195123&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Solubility in Water almost transparency