Octanal ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

O0044-25ML O0044-500ML
TCIAO0044-25MLEA 132 DKK
TCIAO0044-25ML TCIAO0044-500ML
Octanal ≥98.0% (by GC)
Caprylaldehyde
Formel: C₈H₁₆O
Kogepunkt: 170°C
Smeltepunkt: ∼12 °C
Massefylde: 0,82 g/cm³ (20 °C)
Flammepunkt: 52°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007029
CAS-nummer: 124-13-0
EINECS: 204-683-8
UN nummer: 1191
ADR: 3,III
Merck Index: 14,01766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1177e524f30f4fc43612ac1784c9ac1ad6cf274c&vid=44df3ff74d1f23b620ac710679a4c227301326bf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR