3-Chlor-4-Fluorphenylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Chloro-4-fluorobenzeneboronic acid, (3-Chloro-4-fluorophenyl)metaboric acid, Dihydroxy(3-chloro-4-fluorophenyl)borane

C1760-1G C1760-5G
TCIAC1760-1GEA 132 DKK
TCIAC1760-1G TCIAC1760-5G
3-Chlor-4-Fluorphenylboronsyre (contains varying amounts of Anhydride)
3-Chlor-4-Fluorphenylboronsyre
Formel: C₆H₅BClFO₂
MDL Number: MFCD00051800
CAS-nummer: 144432-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e8b57ae5bdb35eb62d185dc2e2dd2c759173cb&vid=39e8b57ae5bdb35eb62d185dc2e2dd2c759173cb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR