D(-)-Ribose ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

R0025-25G R0025-250G
TCIAR0025-25GEA 156 DKK
TCIAR0025-25G TCIAR0025-250G
D(-)-Ribose ≥98.0% (by GC)
D(-)-Ribose
Formel: C₅H₁₀O₅
Kogepunkt: 331 °C (1013 hPa)
Smeltepunkt: 90…95 °C
Massefylde: 0,8 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00135453
CAS-nummer: 50-69-1
EINECS: 200-059-4
Merck Index: 14,08204
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47fb4068d14650f4f7e6b4d08c25bc54826877a2&vid=47fb4068d14650f4f7e6b4d08c25bc54826877a2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -19.0 to -22.0 deg (C=1, H2O)[included NH3 aq. (2+3)]
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR