(R)-(-)-Mandelsyre ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M0662-25G M0662-500G M0662-100G
TCIAM0662-25GEA 279 DKK
TCIAM0662-25G TCIAM0662-500G TCIAM0662-100G
(R)-(-)-Mandelsyre ≥99.0% (by GC, titration analysis)
(R)-(-)-Mandelsyre
Formel: C₈H₈O₃
Kogepunkt: 322 °C (1013 hPa)
Smeltepunkt: 130…132 °C
Massefylde: 1,341 g/cm³ (20 °C)
Flammepunkt: ≥ 190 °C
MDL Number: MFCD00064251
CAS-nummer: 611-71-2
EINECS: 210-276-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=644a09de7bbeb4da749535736d2f891f12d2e57a&vid=644a09de7bbeb4da749535736d2f891f12d2e57a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 99.0 %
Melting point 132.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR