Sorbinsyre ≥99.0% (by titrimetric analysis) for biochemical research
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
S0856-25G
S0856-5G
TCIAS0856-25GEA
230
DKK
TCIAS0856-25G
TCIAS0856-5G
Sorbinsyre ≥99.0% (by titrimetric analysis) for biochemical research
Sorbinsyre
Formel:
CH₃CH=CHCH=CHCOOH Molekylvægt: 112.13 g/mol Kogepunkt: 228°C Flammepunkt: 127 (closed cup) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00002703 CAS-nummer: 110-44-1 EINECS: 203-768-7 Merck Index: 14,08721 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 133.0 to 136.0 deg-C |
Protein impurity test (SDS-PAGE) | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...