L(+)-Arabinos ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A0515-25G
A0515-250G
TCIAA0515-25GEA
271
DKK
TCIAA0515-25G
TCIAA0515-250G
L(+)-Arabinos ≥98.0% (by GC)
L(+)-Arabinos
Formel:
C₅H₁₀O₅ Kogepunkt: 331 °C (1013 hPa) Smeltepunkt: 158…160 °C Massefylde: 1,585 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00135866 CAS-nummer: 5328-37-0 EINECS: 226-214-6 Merck Index: 14,00761 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 152.0 to 160.0 deg-C |
Specific rotation [a]20/D | +102 to +106 deg (C=10, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...