Dihexylsulfid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Di-n-hexylsulphide

H0377-25G H0377-250G
TCIAH0377-25GEA 403 DKK
TCIAH0377-25G TCIAH0377-250G
Dihexylsulfid ≥98.0% (by GC)
Dihexylsulfid
Formel: C₁₂H₂₆S
Molekylvægt: 202.4 g/mol
Kogepunkt: 260°C
Smeltepunkt: -28.3
Flammepunkt: 116°C
MDL Number: MFCD00009528
CAS-nummer: 6294-31-1
EINECS: 228-556-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00151a1512889fbc0ce2678cac17cc2887cc5a13&vid=00151a1512889fbc0ce2678cac17cc2887cc5a13&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR