1-Dodecansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis) for ion pair chromatography

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, Sodium dodecane-1-sulphonate

I0350-25G I0350-5G
TCIAI0350-25GEA 1190 DKK
TCIAI0350-25G TCIAI0350-5G
1-Dodecansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis) for ion pair chromatography
1-Dodecansulfonsyre natriumsalt
Formel: C₁₂H₂₅NaO₃S
Molekylvægt: 272.38 g/mol
Smeltepunkt: 300
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007527
CAS-nummer: 2386-53-0
EINECS: 219-200-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3b0bfbba38897b12a020550698673d3a383ae71&vid=f2a67063561b1cefd87061c26d3fc323c4c28875&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Absorbance (210nm) max. 0.100
Absorbance (230nm) max. 0.030
Absorbance (254nm) max. 0.010


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR