1-Dodecansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, Sodium dodecane-1-sulphonate
D1222-25G
D1222-5G
TCIAD1222-25GEA
1100
DKK
TCIAD1222-25G
TCIAD1222-5G
1-Dodecansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis)
1-Dodecansulfonsyre natriumsalt
Formel:
C₁₂H₂₅NaO₃S Smeltepunkt: 300 °C Massefylde: 1,218 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007527 CAS-nummer: 2386-53-0 EINECS: 219-200-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...