Benzo[a]pyren ≥95.0% (by GC), purified by sublimation

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Benzo[def]chrysen

B0085-1G B0085-100MG
TCIAB0085-1GEA 2370 DKK
TCIAB0085-1G TCIAB0085-100MG
Benzo[a]pyren ≥95.0% (by GC), purified by sublimation
Benzo[a]pyren
Formel: C₂₀H₁₂
Kogepunkt: 496°C
Smeltepunkt: 176…178 °C
Massefylde: 1,351 g/cm³ (20 °C)
MDL Number: MFCD00003602
CAS-nummer: 50-32-8
EINECS: 200-028-5
UN nummer: 3077
ADR: 9,III
Merck Index: 14,01103
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b5174e0a8dd97e7fb65c88d9b178daeadf5dfb7&vid=0f63b2abceee20e85a670a0fe474fdda62b7d317&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 95.0 %
Melting point 176.0 to 180.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR