Pyren ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

P1104-25G P1104-500G P1104-100G
TCIAP1104-25GEA 304 DKK
TCIAP1104-25G TCIAP1104-500G TCIAP1104-100G
Pyren ≥97.0% (by GC)
Pyrene
Formel: C₁₆H₁₀
Molekylvægt: 202.26 g/mol
Kogepunkt: 404°C
MDL Number: MFCD00004136
CAS-nummer: 129-00-0
EINECS: 204-927-3
UN nummer: 2811
ADR: 6.1 II
Merck Index: 14,07963
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87918eb4de97c8725e2efe6b826198a273e81dfe&vid=87918eb4de97c8725e2efe6b826198a273e81dfe&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 148.0 to 152.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR