Inden ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1H-Indene

I0016-25ML I0016-500ML I0016-100ML
TCIAI0016-25MLEA 165 DKK
TCIAI0016-25ML TCIAI0016-500ML TCIAI0016-100ML
Inden ≥93.0% (by GC)
Inden
Formel: C₉H₈
Molekylvægt: 116.16 g/mol
Kogepunkt: 180°C
Smeltepunkt: -2
Flammepunkt: 58°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00003777
CAS-nummer: 95-13-6
EINECS: 202-393-6
UN nummer: 3295
ADR: 3 III
Merck Index: 14,04939
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c3b5c4b4b6de51a3b4cdc2ef5bc29dadf42be77&vid=2c3b5c4b4b6de51a3b4cdc2ef5bc29dadf42be77&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR