Titan(IV)isopropanolat

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T0133-25G T0133-500G
TCIAT0133-25GEA 140 DKK
TCIAT0133-25G TCIAT0133-500G
Titan(IV)isopropanolat
Titan(IV)isopropanolat
Formel: C₁₂H₂₈O₄Ti
Kogepunkt: 220°C
Smeltepunkt: 18…20 °C
Massefylde: 0,9711 g/cm³ (20 °C)
Flammepunkt: 46°C
MDL Number: MFCD00008871
CAS-nummer: 546-68-9
EINECS: 208-909-6
UN nummer: 2413
ADR: 3,III
Merck Index: 14,09480
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3f39390a76ea306ec2c3fa09dd1fb364192d099&vid=a3f39390a76ea306ec2c3fa09dd1fb364192d099&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Ignition residue 27.3 to 28.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR