Aluminiumacetylacetonat ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

A0241-25G A0241-500G A0241-100G
TCIAA0241-25GEA 132 DKK
TCIAA0241-25G TCIAA0241-500G TCIAA0241-100G
Aluminiumacetylacetonat ≥99.0% (by titrimetric analysis)
Aluminiumacetylacetonat
Formel: C₁₅H₂₁AlO₆
Molekylvægt: 324.31 g/mol
Kogepunkt: 315
MDL Number: MFCD00000013
CAS-nummer: 13963-57-0
EINECS: 237-741-6
UN nummer: 3467
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d16ac1d9280e7bba971ad0250f32c827d76dc067&vid=d16ac1d9280e7bba971ad0250f32c827d76dc067&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 99.0 %
Melting point 193.0 to 198.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR