Ceriumtrichlorid, anhydrous ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cerous chloride, Cerium(III) chloride, Cerium (III) chloride

C2058-25G C2058-5G
TCIAC2058-25GEA 1050 DKK
TCIAC2058-25G TCIAC2058-5G
Ceriumtrichlorid, anhydrous ≥98.0% (by titrimetric analysis)
Ceriumtrichlorid
Formel: CeCl₃
Molekylvægt: 246.47 g/mol
Smeltepunkt: 722°C
MDL Number: MFCD00010929
CAS-nummer: 7790-86-5
EINECS: 232-227-8
Merck Index: 14,01997
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e70dcacd02a8624ee52b868b7fa4184277f608c&vid=9c93632eb6cd5d6e1b1083e5e3d96e411c217b07&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR