Natriumtetraphenylborat ≥99.5% (W) precipitation reagent for K

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

A5130-1G A5130-10G
TCIAA5130-1GEA 132 DKK
TCIAA5130-1G TCIAA5130-10G
Natriumtetraphenylborat ≥99.5% (W) precipitation reagent for K
Sodium tetraphenylborate
Formel: (C₆H₅)₄BNa
Molekylvægt: 342.22 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011494
CAS-nummer: 143-66-8
EINECS: 205-605-5
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,08690
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77bb16f6af3a319132328866d2acfd29cf44ae74&vid=db0428e6a501fe5658276754bfe908dba9e54444&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 99.5 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR