General purpose gloves, AlphaTec® 87-190
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
87-190/9.5-10 87-190/6.5-7 87-190/7.5-8 87-190/8.5-9
112-3396EA 12.4 DKK
112-3396 112-3393 112-3394 112-3395
General purpose gloves, AlphaTec® 87-190
Handsker
These natural rubber latex gloves with cotton flock lining provide liquid and mechanical resistance in low risk areas.
  • Specially treated to reduce the risk of allergic reactions
  • Straight cuff
  • Fishscale design provides good grip
They are ideal for use in cleaning, maintenance, light assembly tasks and for janitorial use.
Certificeringer: EN ISO 374-1:2016 Type C, EN ISO 374-5 VIRUS, EN 388:2016 (1010X); Cat. III