Industrial gloves, ActivArmr® 80-100
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80-100/10 80-100/7 80-100/9 80-100/8 80-100/11 80-100/6
ANSE80-100/10EA 510 DKK
ANSE80-100/10 ANSE80-100/7 ANSE80-100/9 ANSE80-100/8 ANSE80-100/11 ANSE80-100/6
Industrial gloves, ActivArmr® 80-100
Handsker
Thanks to its natural rubber coating, the Ansell ActivArmr® 80-100 glove delivers excellent overall mechanical performance for maximal protection and durability.
  • Crinkled finished coating for an excellent dry or wet grip for better safety and productivity
  • Automatic knitted liner delivers an optimally tensioned inner surface for superb fit and flexibility
  • Designed for use in a wide range of handling and carrying applications
Recommended for: Seizing rough blocs and cylinders; seizing and finishing structural parts; loading and unloading rough parts or boxes; picking parts and accessories; final assembly; loading raw material; unloading worked parts.
Certificeringer: EN 388:2016 (2242B); Cat. II