Glove liners, FiberTuf® 76-501
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
76-501/7 76-501/8 76-501/9 76-501/10
112-4361EA 338 DKK
112-4361 112-4362 112-4363 112-4360
Glove liners, FiberTuf® 76-501
Handsker
Made of a new aromatic polyester fibre, FiberTuf® gloves are designed for optimal results in dry applications that require a light mechanical protection, comfort and a good grip.
  • Offer higher performance and durability than traditional cotton, nylon and polyamide gloves
  • Can be used either as primary gloves or undergloves to protect against cold wherever required
  • Dotted PVC coating for good grip
Ideal for general handling, packing, and light assembly applications.
Certificeringer: EN 388:2016 (214XA); Cat. II