Solid phase extraction cartridges, Bond Elut™ Certify
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12102051 12102081 12113050 12113052 12102082 75409050 7540901C 12102085 12102093 75509050 14113050 14113052 14113054 12256145 14102083 14102085 14102093 12256146 14102051 12105030 12102145 12113054 75509025 7550901C 12102051T 12102081T 12256146TJ 14102081T 14113055 52102051 52102081 52113050 52113051
551-0326EA 2860 DKK
551-0326 VARI12102081 551-0350 551-0351 VARI12102082 551-0433 551-0432 VARI12102085 VARI12102093 VARI75509050 VARI14113050 VARI14113052 VARI14113054 VARI12256145 VARI14102083 VARI14102085 VARI14102093 VARI12256146 VARI14102051 VARI12105030 VARI12102145 VARI12113054 VARI75509025 VARI7550901C VARI12102051T VARI12102081T VARI12256146TJ VARI14102081T VARI14113055 VARI52102051 VARI52102081 VARI52113050 VARI52113051
Solid phase extraction cartridges, Bond Elut™ Certify
Kromatografikolonner
The Bond Elut™ Certify extraction cartridge utilises a packed bed consisting of a special, non-polar C8 sorbent and a strong cation exchanger (SCX). Certify is most commonly used to extract basic (cationic) drugs from urine and blood, but it is also very effective for extraction of a wide range of compounds from a diverse range of aqueous matrices.
  • Special mixed mode sorbent bed
  • Broad application range for aqueous extraction
  • Bimodal, non-polar and strong cation exchange