VWR® Autoclavable Bags, 'Sterilbag' and 'Sterilbag indicator'
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
129-0716EA 958 DKK
129-0716 129-0717 119-0057 119-0048
VWR® Autoclavable Bags, 'Sterilbag' and 'Sterilbag indicator'
Poser Affaldsposer
These bags are ideal for autoclaving polystyrene containers and other items contaminated by pathogenic agents (petri dishes, microtitre plates, pipettes, bottles, tubes, etc). They can be used for realising and preserving sterile conditions. Made of special plastic, red coloured.
  • Autoclavable up to 134 °C
  • Available with or without sterilisation indicator strip
  • Red coloured
  • Instructions printed on the bag