Quartz fibre filters
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4170037 4170045 4170050 417009 4170047
MANA4170037EA 3300 DKK
MANA4170037 MANA4170045 MANA4170050 MANA417009 MANA4170047
Quartz fibre filters
Filtre Glasfiberfiltre
Quartz fibre filters feature an extremely low content of metal traces recommended for the analysis of air-borne particles.