Sealable Quartz Glass Cells
Leverandør: HELLMA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
117100F-10-40 117104-10-40 117100-10-40 117104F-10-40 117200-10-40 117200F-10-40 117104F-10K-40
HELL117100F-10-40EA 4960 DKK
HELL117100F-10-40 634-9541 634-9542 HELL117104F-10-40 HELL117200-10-40 HELL117200F-10-40 HELL117104F-10K-40
Sealable Quartz Glass Cells
Kuvetter Spektrofotometerkuvetter
Hellma® high precision quartz glass macro cells for absorption measurements in the UV/Vis range, 10 mm path length, with open screw cap.
  • Cells made of high performance quartz glass
  • Wavelength range from 200 to 2500 nm
  • With open screw cap
  • Silicone seal
  • Volume: 1400 to 3500 μl 
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously