Solid phase extraction cartridges, Bond Elut™ C18 EWP
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12256130 12102136 12102139 12113071 12113068
VARI12256130EA 2810 DKK
VARI12256130 VARI12102136 VARI12102139 VARI12113071 VARI12113068
Solid phase extraction cartridges, Bond Elut™ C18 EWP
Kromatografikolonner
Bond Elut™ EWP is based upon standard particle size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15000 MW), which are typically excluded from standard porosity silica phases.
  • No exclusion of large molecules
  • Good for desalting proteins
  • Successful separation of proteins, peptides or nucleotides