Matrix™ Reagent Reservoirs
Leverandør: THERMO MATRIX TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8086 8095 8096 8093 8094 8085 8075
MATR8086EA 1050 DKK
MATR8086 613-7452 MATR8096 613-7453 613-7451 613-7450 613-5831
Matrix™ Reagent Reservoirs
Reagens reservoirs
PP or PS. These reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills.
  • 75 ml reservoir made of PP is autoclavable and reusable
  • Trough within a trough maximises the amount of liquid accessible to pipette tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduces spillage when pouring reagents out of the reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags so packaging maintains sterility while being easy to open
The 25 ml divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
Emballering: Sterile reservoirs are packaged in snap and tear bags so packaging maintains sterility while being easy to open.