General purpose gloves, Edge® 48-913
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48-913/11 48-913/10 48-913/8 48-913/9
ANSE48-913/11EA 298 DKK
ANSE48-913/11 ANSE48-913/10 ANSE48-913/8 ANSE48-913/9
General purpose gloves, Edge® 48-913
Handsker
These ¾ nitrile coated gloves with cotton jersey lining offer good resistance to abrasive materials.
  • Very flexible
  • Suitable for medium and heavy duty handlings
  • Oil repellent
Ideal for raw material extraction, foundry die casting, primary assembly, warehouse work and repair and maintenance.
Certificeringer: EN 388:2016 (3111B), Cat. II